missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,2-Trichloro-1,1-bis(trifluoromethyl)ethanol, 97%

Manufacturer:  Alfa Aesar™ L1379203

 View more versions of this product

Catalog No. AAL1379203



Specifications

Specifications

2,2,2-Trichloro-1,1-bis(trifluoromethyl)ethanol
1g
C4HCl3F6O
1841641
AMCTWDASUQVRCQ-UHFFFAOYSA-N
1,1,1-trichloro-3,3,3-trifluoro-2-(trifluoromethyl)propan-2-ol
2773244
97%
139°C
7594-49-2
MFCD00236021
1,1-bis trifluoromethyl-2,2,2-trichloroethanol, 1,1,1-trichloro-3,3,3-trifluoro-2-trifluoromethyl propan-2-ol, 2-trichloromethylhexafluoro-2-propanol, 2,2,2-trichloro-1,1-bis trifluoromethyl ethanol, 1,1,1,3,3,3-hexafluoro-2-trichloromethyl-2-propanol, 1,1,1,3,3,3-hexafluoro-2-trichloromethyl propan-2-ol, 1,1,1-trichloro-3,3,3-trifluoro-2-trifluoromethyl-2-propanol, 1,1,1-tris chloranyl-3,3,3-tris fluoranyl-2-trifluoromethyl propan-2-ol
C(C(F)(F)F)(C(F)(F)F)(C(Cl)(Cl)Cl)O
285.391
285.4

Chemical Identifiers

7594-49-2
285.391
AMCTWDASUQVRCQ-UHFFFAOYSA-N
2773244
C(C(F)(F)F)(C(F)(F)F)(C(Cl)(Cl)Cl)O
C4HCl3F6O
MFCD00236021
1,1-bis trifluoromethyl-2,2,2-trichloroethanol, 1,1,1-trichloro-3,3,3-trifluoro-2-trifluoromethyl propan-2-ol, 2-trichloromethylhexafluoro-2-propanol, 2,2,2-trichloro-1,1-bis trifluoromethyl ethanol, 1,1,1,3,3,3-hexafluoro-2-trichloromethyl-2-propanol, 1,1,1,3,3,3-hexafluoro-2-trichloromethyl propan-2-ol, 1,1,1-trichloro-3,3,3-trifluoro-2-trifluoromethyl-2-propanol, 1,1,1-tris chloranyl-3,3,3-tris fluoranyl-2-trifluoromethyl propan-2-ol
1,1,1-trichloro-3,3,3-trifluoro-2-(trifluoromethyl)propan-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : 2° to 8°C

Documents