Please login to your online account to display your discounted pricing

2,2,2-Tribromoethanol, 99%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC421432500

 View more versions of this product

Catalog No. AC421432500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 282.76
Quantity 250g
Beilstein 01, II, 338
Fieser 11,111
Merck Index 12, 9743
CAS Max % 100.0
Packaging Glass bottle
CAS 75-80-9
Assay Percent Range 99%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS Min % 98.5
Linear Formula Br3CCH2OH
Chemical Name or Material 2, 2, 2-Tribromoethanol
MDL Number MFCD00004671
Molecular Formula C2H3Br3O

Specifications

Chemical Name or Material 2, 2, 2-Tribromoethanol
CAS 75-80-9
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C2H3Br3O
Linear Formula Br3CCH2OH
Formula Weight 282.76
MDL Number MFCD00004671
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents