missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[2-(2-Propynyloxy)ethoxy]ethylamine, TCI America™

Manufacturer:  TCI America P22255G

 View more versions of this product

Catalog No. P22255G



Specifications

Specifications

2-[2-(2-Propynyloxy)ethoxy]ethylamine
≥98.0% (GC,T)
2735
5g
GVHLQHHQXWSDDM-UHFFFAOYSA-N
2-(2-prop-2-ynoxyethoxy)ethanamine
55219291
Clear Liquid at 20°C
944561-44-8
C7H13NO2
Yellow
2-[2-(Propargyloxy)ethoxy]ethylamine
C#CCOCCOCCN
143.186
143.19

Chemical Identifiers

944561-44-8
143.186
2-[2-(Propargyloxy)ethoxy]ethylamine
2-(2-prop-2-ynoxyethoxy)ethanamine
C7H13NO2
GVHLQHHQXWSDDM-UHFFFAOYSA-N
55219291
C#CCOCCOCCN
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents