Please login to your online account to display your discounted pricing

2-[(2,2-Dimethylpropanoyl)amino]nicotinic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-[(2, 2-Dimethylpropanoyl)amino]nicotinic acid
CAS 125867-25-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H14N2O3
Formula Weight 222.24
MDL Number MFCD03659710
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00352CB

 
fisher scientific company
MO00352CB
Amber glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 222.24
Quantity 250mg
Assay 97%
CAS Min % 97.0
Packaging Amber glass bottle
Assay Percent Range 97%
CAS 125867-25-6
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-[(2, 2-Dimethylpropanoyl)amino]nicotinic acid
Molecular Formula C11H14N2O3
MDL Number MFCD03659710
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.