Please login to your online account to display your discounted pricing

2-[2-(2-chloroethoxy)ethyl]-1H-isoindole-1,3(2H)-dione, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  KM10098EA

 View more versions of this product

Catalog No. KM10098EA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 253.68
Quantity 10g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
MDL Number MFCD00196075
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-[2-(2-Chloroethoxy)ethyl]-1H-isoindole-1, 3(2H)-dione
Assay Percent Range 97%
CAS 58290-51-0
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C12H12ClNO3
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material 2-[2-(2-Chloroethoxy)ethyl]-1H-isoindole-1, 3(2H)-dione
CAS 58290-51-0
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H12ClNO3
Formula Weight 253.68
MDL Number MFCD00196075
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents