missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[2-(2-chloroethoxy)ethyl]-1H-isoindole-1,3(2H)-dione, 97%, Maybridge

$700.17 - $700.17

Chemical Identifiers

CAS 58290-51-0
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C12H12ClNO3
Formula Weight 253.68
MDL Number MFCD00196075
Chemical Name or Material 2-[2-(2-Chloroethoxy)ethyl]-1H-isoindole-1, 3(2H)-dione
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
KM10098EA SDS Fisher Scientific Company
KM10098EA
Amber Glass Bottle 10g Each for $700.17
Specifications Description & Specifications

Chemical Identifiers

58290-51-0
97%
253.68
2-[2-(2-Chloroethoxy)ethyl]-1H-isoindole-1, 3(2H)-dione
97.0
C12H12ClNO3
MFCD00196075
97%

Specifications

10g
97.0
97%
253.68
2-[2-(2-Chloroethoxy)ethyl]-1H-isoindole-1, 3(2H)-dione
58290-51-0
Amber Glass Bottle
C12H12ClNO3
MFCD00196075
97%

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger