missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,2,4′-Tetrafluoroacetophenone, TCI America™

Manufacturer:  TCI America T325725G

 View more versions of this product

Catalog No. T325725G



Specifications

Specifications

2,2,2,4′-Tetrafluoroacetophenone
White-Yellow
655-32-3
MFCD00042393
LUKLMXJAEKXROG-UHFFFAOYSA-N
2,2,2-trifluoro-1-(4-fluorophenyl)ethanone
69564
Crystal-Lump at 20°C
152°C
25g
C8H4F4O
1325
C1=CC(=CC=C1C(=O)C(F)(F)F)F
192.113
192.11
≥98.0% (GC)

Chemical Identifiers

655-32-3
192.113
LUKLMXJAEKXROG-UHFFFAOYSA-N
2,2,2-trifluoro-1-(4-fluorophenyl)ethanone
C8H4F4O
MFCD00042393
69564
C1=CC(=CC=C1C(=O)C(F)(F)F)F
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

Documents