Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2,2,2-Trimethylacetamido)benzeneboronic acid, 95%

$99.14 - $219.99

Chemical Identifiers

Chemical Name or Material 2-(2,2,2-Trimethylacetamido)benzeneboronic acid
CAS 146140-95-6
Molecular Formula C11H16BNO3
Formula Weight 221.06
MDL Number MFCD01114645
Synonym 2-Pivalamidophenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2700403

  SDS
alfa aesar™
H2700403
1g Each for $219.99

AAH27004MD

  SDS
alfa aesar™
H27004MD
250mg Each for $99.14
Description & Specifications

Specifications

Formula Weight 221.06
Chemical Name or Material 2-(2,2,2-Trimethylacetamido)benzeneboronic acid
Assay 95%
Molecular Formula C11H16BNO3
MDL Number MFCD01114645
CAS 146140-95-6
Synonym 2-Pivalamidophenylboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 268° to 271°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.