Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2,2,2-Trifluoroethoxy)pyridine-3-carboxaldehyde, 96%

$1,074.32 - $1,074.32

Chemical Identifiers

Chemical Name or Material 2-(2,2,2-Trifluoroethoxy)pyridine-3-carboxaldehyde
CAS 902837-51-8
Molecular Formula C8H6F3NO2
Formula Weight 205.14
MDL Number MFCD07781175
Synonym 2-(2,2,2-Trifluoroethoxy)nicotinaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5011806

  SDS
alfa aesar™
H5011806
5g Each for $1,074.32
Description & Specifications

Specifications

Formula Weight 205.14
Chemical Name or Material 2-(2,2,2-Trifluoroethoxy)pyridine-3-carboxaldehyde
Assay 96%
Molecular Formula C8H6F3NO2
MDL Number MFCD07781175
CAS 902837-51-8
Synonym 2-(2,2,2-Trifluoroethoxy)nicotinaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.