Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(1H-Tetrazol-5-yl)phenol, 97%

$258.59 - $781.96

Chemical Identifiers

Chemical Name or Material 2-(1H-Tetrazol-5-yl)phenol
CAS 51449-77-5
Molecular Formula C7H6N4O
Formula Weight 162.16
MDL Number MFCD09037787
Synonym 5-(2-Hydroxyphenyl)-1H-tetrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2767203

 
alfa aesar™
H2767203
1g Each for $258.59

AAH2767206

 
alfa aesar™
H2767206
5g Each for $781.96
Description & Specifications

Specifications

Formula Weight 162.16
Chemical Name or Material 2-(1H-Tetrazol-5-yl)phenol
Assay 97%
Molecular Formula C7H6N4O
MDL Number MFCD09037787
CAS 51449-77-5
Synonym 5-(2-Hydroxyphenyl)-1H-tetrazole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈230°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.