missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2(1H)-Quinolinone-3-carboxaldehyde, 97%

Manufacturer:  Alfa Aesar™ H3350803

 View more versions of this product

Catalog No. AAH3350803



Specifications

Specifications

2(1H)-Quinolinone-3-carboxaldehyde
White to Brown
1g
C10H7NO2
2-hydroxyquinoline-3-carbaldehyde, 2-oxo-1,2-dihydro-quinoline-3-carbaldehyde, 2-oxo-1,2-dihydroquinoline-3-carbaldehyde, 3-formyl-2quinolone, 1,2-dihydro-2-oxoquinoline-3-carboxaldehyde, 1,2-dihydro-2-oxoquinoline-3-carbaldehyde, 2-hydroxy-3-quinolinecarboxaldehyde, 2-oxo-1,2-dihydro-3-quinolinecarbaldehyde, 2-oxohydroquinoline-3-carbaldehyde, 3-formylcarbostyril
C1=CC=C2C(=C1)C=C(C(=O)N2)C=O
173.171
173.2
97%
Air Sensitive
>250°C
91301-03-0
MFCD00563443
VWHKEYXRRNSJTN-UHFFFAOYSA-N
2-oxo-1H-quinoline-3-carbaldehyde
589334
Solid

Chemical Identifiers

91301-03-0
173.171
VWHKEYXRRNSJTN-UHFFFAOYSA-N
589334
C1=CC=C2C(=C1)C=C(C(=O)N2)C=O
C10H7NO2
MFCD00563443
2-hydroxyquinoline-3-carbaldehyde, 2-oxo-1,2-dihydro-quinoline-3-carbaldehyde, 2-oxo-1,2-dihydroquinoline-3-carbaldehyde, 3-formyl-2quinolone, 1,2-dihydro-2-oxoquinoline-3-carboxaldehyde, 1,2-dihydro-2-oxoquinoline-3-carbaldehyde, 2-hydroxy-3-quinolinecarboxaldehyde, 2-oxo-1,2-dihydro-3-quinolinecarbaldehyde, 2-oxohydroquinoline-3-carbaldehyde, 3-formylcarbostyril
2-oxo-1H-quinoline-3-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents