Please login to your online account to display your discounted pricing

Alfa Aesar™ 2(1H)-Quinolinone-3-carboxaldehyde, 97%

Chemical Identifiers

Chemical Name or Material 2(1H)-Quinolinone-3-carboxaldehyde
CAS 91301-03-0
Formula Weight 173.2
MDL Number MFCD00563443
Synonym 2-Hydroxyquinoline-3-carboxaldehyde; 2-Oxo-1,2-dihydroquinoline-3-carboxaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3350803

  SDS
alfa aesar™
H3350803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3350806

  SDS
alfa aesar™
H3350806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH33508MD

  SDS
alfa aesar™
H33508MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 173.2
Chemical Name or Material 2(1H)-Quinolinone-3-carboxaldehyde
Physical Form Solid
Assay 97%
MDL Number MFCD00563443
CAS 91301-03-0
Synonym 2-Hydroxyquinoline-3-carboxaldehyde; 2-Oxo-1,2-dihydroquinoline-3-carboxaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Air Sensitive
Color White to brown
Melting Point >250°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.