missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(1H-Pyrrol-1-yl)benzoic acid, ≥97%, Maybridge

Manufacturer:  Fisher Scientific Company CC34301DE

 View more versions of this product

Catalog No. CC34301DE

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

187.2
97%
MFCD00051645
97%
96.5
5g
100.0
10333-68-3
2-(1H-Pyrrol-1-yl)benzoic acid
C11H9NO2

Chemical Identifiers

10333-68-3
100.0
C11H9NO2
MFCD00051645
97%
96.5
97%
187.2
2-(1H-Pyrrol-1-yl)benzoic acid

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.