missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(1H-Pyrrol-1-yl)benzoic acid, ≥97%, Maybridge

Chemical Identifiers

CAS 10333-68-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H9NO2
Formula Weight 187.2
MDL Number MFCD00051645
Chemical Name or Material 2-(1H-Pyrrol-1-yl)benzoic acid
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC34301DE Fisher Scientific Company
CC34301DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC34301DA SDS Fisher Scientific Company
CC34301DA
Amber glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

10333-68-3
100.0
C11H9NO2
MFCD00051645
97%
96.5
97%
187.2
2-(1H-Pyrrol-1-yl)benzoic acid

Specifications

187.2
100.0
10333-68-3
96.5
2-(1H-Pyrrol-1-yl)benzoic acid
5g
MFCD00051645
97%
C11H9NO2
97%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.