missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(1H-Pyrazol-3-yl)phenol, TCI America™

Manufacturer:  TCI America P22185G

 View more versions of this product

Catalog No. P22185G



Specifications

Specifications

2-(1H-Pyrazol-3-yl)phenol
97°C
34810-67-8
C9H8N2O
3-(2-Hydroxyphenyl)pyrazole
C1=CC(=C2C=CNN2)C(=O)C=C1
160.176
160.18
White-Yellow
5g
≥98.0% (GC,T)
MFCD03093876
SUWASPSYOCZZQV-UHFFFAOYSA-N
6-(1,2-dihydropyrazol-3-ylidene)cyclohexa-2,4-dien-1-one
6743769
Crystal-Powder at 20°C

Chemical Identifiers

34810-67-8
160.176
SUWASPSYOCZZQV-UHFFFAOYSA-N
6743769
C1=CC(=C2C=CNN2)C(=O)C=C1
C9H8N2O
MFCD03093876
3-(2-Hydroxyphenyl)pyrazole
6-(1,2-dihydropyrazol-3-ylidene)cyclohexa-2,4-dien-1-one
Safety and Handling

Safety and Handling

TSCA : No

Documents