missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(1H-Pyrazol-1-yl)benzaldehyde, ≥97%, Maybridge

Chemical Identifiers

CAS 138479-47-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H8N2O
Formula Weight 172.19
MDL Number MFCD03086186
Chemical Name or Material 2-(1H-Pyrazol-1-yl)benzaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC24404CB SDS Fisher Scientific Company
CC24404CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

138479-47-7
100.0
C10H8N2O
MFCD03086186
97%
96.5
97%
172.19
2-(1H-Pyrazol-1-yl)benzaldehyde

Specifications

250mg
96.5
97%
172.19
2-(1H-Pyrazol-1-yl)benzaldehyde
138479-47-7
100.0
C10H8N2O
MFCD03086186
97%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.