Please login to your online account to display your discounted pricing

2-(1H-indol-3-yl)ethan-1-ol, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(1H-Indol-3-yl)ethan-1-ol
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H11NO
Formula Weight 161.2
Synonym β-3-Indolylethanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB14321EA

  SDS
fisher scientific company
BTB14321EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB14321FA

 
fisher scientific company
BTB14321FA
Amber glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB14321EE

  SDS
fisher scientific company
BTB14321EE
Amber Glass Bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 161.2
Quantity 10g
Assay 97%
Merck Index 15, 9978
Packaging Amber Glass Bottle
Synonym β-3-Indolylethanol
Molecular Formula C10H11NO
Assay Percent Range 97%
CAS 526-55-6
Chemical Name or Material 2-(1H-Indol-3-yl)ethan-1-ol