Please login to your online account to display your discounted pricing

2-(1H-indol-3-yl)ethan-1-ol, Maybridge


Quantity 50g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB14321EE

 View more versions of this product

Catalog No. BTB14321EE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 161.2
Quantity 50g
Merck Index 15, 9978
Assay 97%
Packaging Amber Glass Bottle
Molecular Formula C10H11NO
CAS 526-55-6
Chemical Name or Material 2-(1H-Indol-3-yl)ethan-1-ol
Assay Percent Range 97%
Synonym β-3-Indolylethanol

Specifications

Chemical Name or Material 2-(1H-Indol-3-yl)ethan-1-ol
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H11NO
Formula Weight 161.2
Synonym β-3-Indolylethanol