missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(1H-Imidazol-1-yl)benzylamine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444090010
Specifications
2-(1H-Imidazol-1-yl)benzylamine | |
Glass Bottle | |
Yellow | |
1g | |
96% min. (HPLC) | |
1-2-1h-imidazol-1-yl phenyl methanamine, 1-2-imidazol-1-yl phenyl methanamine, 2-1h-imidazol-1-yl benzylamine, 2-1h-imidazol-1-yl phenyl methanamine, 2-imidazol-1-yl phenyl methanamine, 2-imidazol-1-yl-benzylamine, 2-imidazol-1-ylbenzylamine, 2-imidazolylphenyl methylamine, benzenemethanamine, 2-1h-imidazol-1-yl | |
C1=CC=C(C(=C1)CN)N2C=CN=C2 | |
173.22 | |
173.22 | |
97% |
Authentic | |
180°C to 200°C (2.5mmHg) | |
51°C to 52°C | |
25373-55-1 | |
C10H11N3 | |
JHDMBRSDCFLSTD-UHFFFAOYSA-N | |
(2-imidazol-1-ylphenyl)methanamine | |
3161671 | |
Crystalline Powder |
Chemical Identifiers
25373-55-1 | |
173.22 | |
1-2-1h-imidazol-1-yl phenyl methanamine, 1-2-imidazol-1-yl phenyl methanamine, 2-1h-imidazol-1-yl benzylamine, 2-1h-imidazol-1-yl phenyl methanamine, 2-imidazol-1-yl phenyl methanamine, 2-imidazol-1-yl-benzylamine, 2-imidazol-1-ylbenzylamine, 2-imidazolylphenyl methylamine, benzenemethanamine, 2-1h-imidazol-1-yl | |
(2-imidazol-1-ylphenyl)methanamine |
C10H11N3 | |
JHDMBRSDCFLSTD-UHFFFAOYSA-N | |
3161671 | |
C1=CC=C(C(=C1)CN)N2C=CN=C2 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger