missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(1H-Imidazol-1-yl)benzylamine, 97%, ACROS Organics™

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C10H11N3
Formula Weight 173.22
Synonym 1-(α-Amino-o-tolyl)imidazole
Chemical Name or Material 2-(1H-Imidazol-1-yl)benzylamine
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC444090010 Thermo Fisher Scientific
AC444090010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC444090050 Thermo Fisher Scientific
AC444090050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
173.22
2-(1H-Imidazol-1-yl)benzylamine
C10H11N3
1-(α-Amino-o-tolyl)imidazole
≥96.0%

Specifications

25373-55-1
Crystalline Powder
C10H11N3
Pale yellow
1-(α-Amino-o-tolyl)imidazole
2-(1H-Imidazol-1-yl)benzylamine
Authentic
97%
173.22
50° to 54°C
1g
≥96.0%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger