missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(1H-1,2,4-Triazol-3-yl)pyridine, TCI America™

Manufacturer:  TCI America P21001G

 View more versions of this product

Catalog No. P21001G



Specifications

Specifications

2-(1H-1,2,4-Triazol-3-yl)pyridine
162°C
23195-62-2
MFCD13188589
PVMLJDLTWMGZAH-UHFFFAOYSA-N
2-(1H-1,2,4-triazol-5-yl)pyridine
10898842
Crystal-Powder at 20°C
White-Yellow
1g
C7H6N4
3-(2-Pyridyl)-1,2,4-triazole
C1=CC=NC(=C1)C2=NC=NN2
146.153
146.15
≥98.0% (GC,T)

Chemical Identifiers

23195-62-2
146.153
PVMLJDLTWMGZAH-UHFFFAOYSA-N
10898842
C1=CC=NC(=C1)C2=NC=NN2
C7H6N4
MFCD13188589
3-(2-Pyridyl)-1,2,4-triazole
2-(1H-1,2,4-triazol-5-yl)pyridine
Safety and Handling

Safety and Handling

TSCA : No

Documents