missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(1H-1,2,4-triazol-1-ylmethyl)benzoic acid, 97%, Maybridge

Chemical Identifiers

CAS 876718-01-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H9N3O2
Formula Weight 203.2
MDL Number MFCD07186455
Chemical Name or Material 2-(1H-1, 2, 4-Triazol-1-ylmethyl)benzoic acid
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC47501DA SDS Fisher Scientific Company
CC47501DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

876718-01-3
100.0
C10H9N3O2
MFCD07186455
97%
96.5
97%
203.2
2-(1H-1, 2, 4-Triazol-1-ylmethyl)benzoic acid

Specifications

203.2
100.0
C10H9N3O2
96.5
97%
97%
1g
MFCD07186455
876718-01-3
Amber Glass Bottle
2-(1H-1, 2, 4-Triazol-1-ylmethyl)benzoic acid

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning