Please login to your online account to display your discounted pricing

2-[[(1E)-2-(Methoxycarbonyl)-1-methylvinyl]amino]-(2R)-2-cyclohexa-1,4-dienylacetic acid, sodium salt, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  335990010

 View more versions of this product

Catalog No. AC335990010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 273.26
Quantity 1g
TSCA TSCA
EINECS Number 288-783-7
CAS Max % 100.0
CAS Min % 97.5
MDL Number MFCD00196811
Packaging Glass bottle
Name Note Na salt, 98%
Molecular Formula C13H16NNaO4
CAS 85896-06-6
Chemical Name or Material 2-[[(1E)-2-(Methoxycarbonyl)-1-methylvinyl]amino]-(2R)-2-cyclohexa-1, 4-dienylacetic acid, sodium salt
Linear Formula C6H7CH(COONa)NHC(CH3)CHC(O)OCH3
Assay Percent Range 98%

Specifications

Chemical Name or Material 2-[[(1E)-2-(Methoxycarbonyl)-1-methylvinyl]amino]-(2R)-2-cyclohexa-1, 4-dienylacetic acid, sodium salt
Name Note Na salt, 98%
CAS 85896-06-6
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C13H16NNaO4
Linear Formula C6H7CH(COONa)NHC(CH3)CHC(O)OCH3
Formula Weight 273.26
MDL Number MFCD00196811