Please login to your online account to display your discounted pricing

2-[[(1E)-2-(Methoxycarbonyl)-1-methylvinyl]amino]-(2R)-2-cyclohexa-1,4-dienylacetic acid, sodium salt, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-[[(1E)-2-(Methoxycarbonyl)-1-methylvinyl]amino]-(2R)-2-cyclohexa-1, 4-dienylacetic acid, sodium salt
Name Note Na salt, 98%
CAS 85896-06-6
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C13H16NNaO4
Linear Formula C6H7CH(COONa)NHC(CH3)CHC(O)OCH3
Formula Weight 273.26
MDL Number MFCD00196811
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC335990010

  SDS
thermo fisher scientific
335990010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC335990050

  SDS
thermo fisher scientific
335990050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 273.26
Quantity 1g
TSCA TSCA
EINECS Number 288-783-7
CAS Max % 100.0
CAS Min % 97.5
MDL Number MFCD00196811
Packaging Glass bottle
Name Note Na salt, 98%
Molecular Formula C13H16NNaO4
CAS 85896-06-6
Chemical Name or Material 2-[[(1E)-2-(Methoxycarbonyl)-1-methylvinyl]amino]-(2R)-2-cyclohexa-1, 4-dienylacetic acid, sodium salt
Linear Formula C6H7CH(COONa)NHC(CH3)CHC(O)OCH3
Assay Percent Range 98%