missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[(11bR)-3H-Dinaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylammonium tetrafluoroborate, 97+%, Alfa Aesar™

Chemical Identifiers

CAS 1223007-01-9
Molecular Formula C24H23BF4NP
Molecular Weight (g/mol) 443.232
MDL Number MFCD17018773
InChI Key AMEHGPFCNMMXTB-UHFFFAOYSA-O
Synonym 2-11br-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11bs-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11br-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-11bs-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 15 ,2,4,6,8,10,16,18,20,22-decaen-13-yl ethanaminium tetrafluoroborate
PubChem CID 72376450
SMILES [B-](F)(F)(F)F.C1C2=C(C3=CC=CC=C3C=C2)C4=C(CP1CC[NH3+])C=CC5=CC=CC=C54
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60215MD View Documents Alfa Aesar
H60215MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6021503 View Documents Alfa Aesar
H6021503
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1223007-01-9
443.232
AMEHGPFCNMMXTB-UHFFFAOYSA-O
72376450
C24H23BF4NP
MFCD17018773
2-11br-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11bs-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11br-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-11bs-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 15 ,2,4,6,8,10,16,18,20,22-decaen-13-yl ethanaminium tetrafluoroborate
[B-](F)(F)(F)F.C1C2=C(C3=CC=CC=C3C=C2)C4=C(CP1CC[NH3+])C=CC5=CC=CC=C54

Specifications

250mg
C24H23BF4NP
2-11br-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11bs-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11br-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-11bs-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 15 ,2,4,6,8,10,16,18,20,22-decaen-13-yl ethanaminium tetrafluoroborate
[B-](F)(F)(F)F.C1C2=C(C3=CC=CC=C3C=C2)C4=C(CP1CC[NH3+])C=CC5=CC=CC=C54
72376450
≥97%
1223007-01-9
MFCD17018773
AMEHGPFCNMMXTB-UHFFFAOYSA-O
443.232
443.22
2-[(11bR)-3H-Dinaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylammonium tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning