Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,10-Dodecadiyne, 97%


Quantity 5g

Manufacturer: alfa aesar™  L1282206

 View more versions of this product

Catalog No. AAL1282206

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 162.28
Chemical Name or Material 2,10-Dodecadiyne
MDL Number MFCD00041622
CAS 31699-38-4
Molecular Formula C12H18
Assay 97%
Boiling Point 103° to 104°C (8mmHg)
Quantity 5g

Specifications

Chemical Name or Material 2,10-Dodecadiyne
CAS 31699-38-4
Molecular Formula C12H18
Formula Weight 162.28
MDL Number MFCD00041622