Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,10-Dodecadiyne, 97%

$54.39 - $180.81

Chemical Identifiers

Chemical Name or Material 2,10-Dodecadiyne
CAS 31699-38-4
Molecular Formula C12H18
Formula Weight 162.28
MDL Number MFCD00041622
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1282206

  SDS
alfa aesar™
L1282206
5g Each for $54.39

AAL1282214

  SDS
alfa aesar™
L1282214
25g Each for $180.81
Description & Specifications

Specifications

Formula Weight 162.28
Chemical Name or Material 2,10-Dodecadiyne
MDL Number MFCD00041622
CAS 31699-38-4
Molecular Formula C12H18
Assay 97%
Boiling Point 103° to 104°C (8mmHg)
Quantity 5g