Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,10-Dodecadiyne, 97%


Quantity 25g

Manufacturer: alfa aesar™  L1282214

 View more versions of this product

Catalog No. AAL1282214

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 162.28
CAS 31699-38-4
Assay 97%
Molecular Formula C12H18
MDL Number MFCD00041622
Boiling Point 103° to 104°C (8mmHg)
Quantity 25g
Chemical Name or Material 2,10-Dodecadiyne

Specifications

Chemical Name or Material 2,10-Dodecadiyne
CAS 31699-38-4
Molecular Formula C12H18
Formula Weight 162.28
MDL Number MFCD00041622