Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester, 95%

$197.39 - $197.39

Chemical Identifiers

Chemical Name or Material 2-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester
CAS 1073354-41-2
Molecular Formula C15H23BN2O2
Formula Weight 274.17
MDL Number MFCD06798280
Synonym 2-(1-Pyrrolidinyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine; 2-(1-Pyrrolidino)pyridine-3-boronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27190MD

  SDS
alfa aesar™
H27190MD
250mg Each for $197.39
Description & Specifications

Specifications

Formula Weight 274.17
MDL Number MFCD06798280
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester
Molecular Formula C15H23BN2O2
Health Hazard 1 Warning
CAS 1073354-41-2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
Melting Point 76° to 78°C
Quantity 250mg
Synonym 2-(1-Pyrrolidinyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine; 2-(1-Pyrrolidino)pyridine-3-boronic acid pinacol ester

Applications
2-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.