Learn More
2-(1-Piperidinyl)phenol, 98%, Thermo Scientific Chemicals
$72.95 - $262.48
Chemical Identifiers
CAS | 65195-20-2 |
---|---|
Molecular Formula | C11H15NO |
Molecular Weight (g/mol) | 177.25 |
MDL Number | MFCD00051687 |
InChI Key | OXELIFPFCCGAJX-UHFFFAOYSA-N |
Synonym | 2-piperidin-1-yl phenol, 2-piperidinophenol, 2-1-piperidino phenol, 2-1-piperidinyl phenol, 2-piperidylphenol, 2-hydroxyphenyl piperidine, 1-2-hydroxyphenyl piperidine |
PubChem CID | 3262250 |
SMILES | OC1=CC=CC=C1N1CCCCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0763003
|
Thermo Scientific Chemicals
L0763003 |
1 g |
Each for $72.95
|
|
|||||
AAL0763006
|
Thermo Scientific Chemicals
L0763006 |
5 g |
Each for $262.48
|
|
|||||
Description
2-(1-Piperidinyl)phenol is used as primary and secondary intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(1-Piperidinyl)phenol is used as primary and secondary intermediates.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents.
Chemical Identifiers
65195-20-2 | |
177.25 | |
OXELIFPFCCGAJX-UHFFFAOYSA-N | |
3262250 |
C11H15NO | |
MFCD00051687 | |
2-piperidin-1-yl phenol, 2-piperidinophenol, 2-1-piperidino phenol, 2-1-piperidinyl phenol, 2-piperidylphenol, 2-hydroxyphenyl piperidine, 1-2-hydroxyphenyl piperidine | |
OC1=CC=CC=C1N1CCCCC1 |
Specifications
65195-20-2 | |
C11H15NO | |
1 g | |
2-piperidin-1-yl phenol, 2-piperidinophenol, 2-1-piperidino phenol, 2-1-piperidinyl phenol, 2-piperidylphenol, 2-hydroxyphenyl piperidine, 1-2-hydroxyphenyl piperidine | |
OC1=CC=CC=C1N1CCCCC1 | |
177.25 | |
177.24 | |
2-(1-Piperidinyl)phenol |
73°C to 76°C | |
MFCD00051687 | |
151777 | |
OXELIFPFCCGAJX-UHFFFAOYSA-N | |
2-(piperidin-1-yl)phenol | |
3262250 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only