Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(1-Piperidinyl)ethyl isothiocyanate, 97%

Chemical Identifiers

Chemical Name or Material 2-(1-Piperidinyl)ethyl isothiocyanate
CAS 32813-24-4
Molecular Formula C8H14N2S
Formula Weight 170.28
MDL Number MFCD00041232
UN Number UN2922
Beilstein 1238446
Synonym 1-(2-Isothiocyanatoethyl)piperidine; 2-(1-Piperidino)ethyl isothiocyanate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0946303

  SDS
alfa aesar™
L0946303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0946306

  SDS
alfa aesar™
L0946306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 170.28
MDL Number MFCD00041232
Assay 97%
Molecular Formula C8H14N2S
CAS 32813-24-4
Synonym 1-(2-Isothiocyanatoethyl)piperidine; 2-(1-Piperidino)ethyl isothiocyanate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Boiling Point 84°C (5mmHg)
Quantity 1g
Chemical Name or Material 2-(1-Piperidinyl)ethyl isothiocyanate
UN Number UN2922
Beilstein 1238446
Flash Point >110°C (230°F)
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

2° to 8°C