missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(1-Piperidinyl)benzonitrile, 97%

Manufacturer:  Alfa Aesar™ A1178403

 View more versions of this product

Catalog No. AAA1178403


 

Specifications

Specifications

2- (1-Piperidinyl)benzonitrile
41° to 45°C
72752-52-4
C12H14N2
UN3077
1-(2-Cyanophenyl)piperidine; 2-(1-Piperidino)benzonitrile
C1CCN(CC1)C2=CC=CC=C2C#N
186.258
186.26
122° to 125°C (1mmHg)
1g
97%
MFCD00049221
1243909
MEBVSLLKZSAIGK-UHFFFAOYSA-N
2-piperidin-1-ylbenzonitrile
2774355

Chemical Identifiers

72752-52-4
186.258
MEBVSLLKZSAIGK-UHFFFAOYSA-N
2774355
C12H14N2
MFCD00049221
1-(2-Cyanophenyl)piperidine; 2-(1-Piperidino)benzonitrile
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

SDS Documents