Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(1-Piperidinyl)acetamidoxime, 97%

$657.27 - $657.27

Chemical Identifiers

Chemical Name or Material 2-(1-Piperidinyl)acetamidoxime
CAS 175136-64-8
Molecular Formula C7H15N3O
Formula Weight 157.22g/mol
MDL Number MFCD00052662
Synonym N-Hydroxy-1-piperidineethanimidamide; 2-(1-Piperidino)acetamidoxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5065606

 
alfa aesar™
H50656-06
5g Each for $657.27
Description & Specifications

Specifications

Quantity 5g
Quantity 5g
Chemical Name or Material 2-(1-Piperidinyl)acetamidoxime
CAS 175136-64-8
Assay 97%
Molecular Formula C7H15N3O
Formula Weight 157.22g/mol
MDL Number MFCD00052662
Synonym N-Hydroxy-1-piperidineethanimidamide; 2-(1-Piperidino)acetamidoxime
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.