Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(1-Piperidinyl)-5-(trifluoromethyl)aniline

Chemical Identifiers

Chemical Name or Material 2-(1-Piperidinyl)-5-(trifluoromethyl)aniline
CAS 1496-40-8
Molecular Formula C12H15F3N2
Formula Weight 244.26
MDL Number MFCD00042161
UN Number UN2811
Synonym 3-Amino-4-(1-piperidinyl)benzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5072303

  SDS
alfa aesar™
H5072303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5072306

  SDS
alfa aesar™
H5072306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 244.26
Chemical Name or Material 2-(1-Piperidinyl)-5-(trifluoromethyl)aniline
Molecular Formula C12H15F3N2
MDL Number MFCD00042161
CAS 1496-40-8
UN Number UN2811
Synonym 3-Amino-4-(1-piperidinyl)benzotrifluoride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.