Please login to your online account to display your discounted pricing

2-(1-Piperidino)aniline, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(1-Piperidino)aniline
Name Note 98%
CAS 39643-31-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H16N2
Formula Weight 176.26
MDL Number MFCD00047467
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC369250010

  SDS
fisher scientific acros
AC369250010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC369250050

  SDS
thermo fisher scientific
AC369250050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystals or Crystalline Powder
Formula Weight 176.26
Quantity 1g
Color Brown
Melting Point 44° to 48°C
CAS Max % 100.0
Name Note 98%
Molecular Formula C11H16N2
Health Hazard 1 GHS Signal Word: Warning
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-(1-Piperidino)aniline
Assay Percent Range 98%
MDL Number MFCD00047467
CAS 39643-31-7
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Packaging Glass bottle
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.