missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(1-Piperazinyl)pyrimidine dihydrochloride, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC243290250

 View more versions of this product

Catalog No. AC243290250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-(1-Piperazinyl)pyrimidine dihydrochloride
Glass bottle
286.0°C to 288.0°C
94021-22-4
100.0
C8H12N4·2HCl
1-2-pyrimidinyl piperazine dihydrochloride, 1-2-pyrimidyl piperazine dihydrochloride, 2-1-piperazinyl pyrimidine dihydrochloride, 2-piperazin-1-yl pyrimidine dihydrochloride, 2-piperazin-1-ylpyrimidine dihydrochloride, 2-piperazinylpyrimidine dihydrochloride, 2-piperazinylpyrimidine hcl, pubchem7349, pyrimidine, 2-1-piperazinyl-, dihydrochloride, pyrimidinylpiperazine dihydrochloride
C1CN(CCN1)C2=NC=CC=N2.Cl.Cl
237.13
237.13
98+%
Authentic
Beige to White
25g
98.0
98+%
MFCD00012779
ZNZGJSLHXOMREP-UHFFFAOYSA-N
2-piperazin-1-ylpyrimidine;dihydrochloride
11680272
Powder

Chemical Identifiers

94021-22-4
237.13
ZNZGJSLHXOMREP-UHFFFAOYSA-N
11680272
C1CN(CCN1)C2=NC=CC=N2.Cl.Cl
C8H12N4·2HCl
MFCD00012779
1-2-pyrimidinyl piperazine dihydrochloride, 1-2-pyrimidyl piperazine dihydrochloride, 2-1-piperazinyl pyrimidine dihydrochloride, 2-piperazin-1-yl pyrimidine dihydrochloride, 2-piperazin-1-ylpyrimidine dihydrochloride, 2-piperazinylpyrimidine dihydrochloride, 2-piperazinylpyrimidine hcl, pubchem7349, pyrimidine, 2-1-piperazinyl-, dihydrochloride, pyrimidinylpiperazine dihydrochloride
2-piperazin-1-ylpyrimidine;dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 301-415-2

SDS Documents