Please login to your online account to display your discounted pricing

2-(1-Piperazinyl)pyrimidine dihydrochloride, 98+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  243290050

 View more versions of this product

Catalog No. AC243290050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Powder
Formula Weight 237.13
Quantity 5g
Color White to very slightly beige
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material 2-(1-Piperazinyl)pyrimidine dihydrochloride
CAS 94021-22-4
Assay Percent Range 98+%
Molecular Formula C8H12N4·2HCl
MDL Number MFCD00012779
Synonym 1-(2-Pyrimidyl)piperazine dihydrochloride
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material 2-(1-Piperazinyl)pyrimidine dihydrochloride
CAS 94021-22-4
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C8H12N4·2HCl
Formula Weight 237.13
MDL Number MFCD00012779
Synonym 1-(2-Pyrimidyl)piperazine dihydrochloride
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents