missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(1-Piperazinyl)pyrimidine, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC134320250

 View more versions of this product

Catalog No. AC134320250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

164.21
≥98.0%
Clear yellow
25, IV, 2083
98.0
Glass bottle
20980-22-7
MFCD00040742
1-(2-Pyrimidyl)piperazine
Liquid After Melting
25g
2-(1-Piperazinyl)pyrimidine
100.0
1.5850 to 1.5880 (20°C, 589nm)
C8H12N4
2-Piperazinopyrimidine
Authentic
98+%

Chemical Identifiers

20980-22-7
100.0
C8H12N4
MFCD00040742
≥98.0%
98.0
98+%
164.21
2-Piperazinopyrimidine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger