Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(1-Piperazinyl)nicotinamide, 95%

$326.32 - $326.32

Chemical Identifiers

Chemical Name or Material 2-(1-Piperazinyl)nicotinamide
CAS 87394-64-7
Molecular Formula C10H14N4O
Formula Weight 206.25
MDL Number MFCD08061093
Synonym 2-(1-Piperazino)nicotinamide; 2-(1-Piperazinyl)pyridine-3-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5096703

 
alfa aesar™
H5096703
1g Each for $326.32
Description & Specifications

Specifications

Formula Weight 206.25
Chemical Name or Material 2-(1-Piperazinyl)nicotinamide
Assay 95%
Molecular Formula C10H14N4O
MDL Number MFCD08061093
CAS 87394-64-7
Synonym 2-(1-Piperazino)nicotinamide; 2-(1-Piperazinyl)pyridine-3-carboxamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.