Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(1-Naphthylmethyl)-2-imidazoline nitrate, 99%

$59.22 - $173.28

Chemical Identifiers

Chemical Name or Material 2-(1-Naphthylmethyl)-2-imidazoline nitrate
CAS 5144-52-5
Molecular Formula C14H14N2·HNO3
Formula Weight 273.29
MDL Number MFCD00014316
Beilstein 3779329
Synonym Naphazoline nitrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1261714

  SDS
alfa aesar™
L1261714
25g Each for $59.22

AAL1261722

  SDS
alfa aesar™
L1261722
100g Each for $173.28
Description & Specifications

Specifications

Formula Weight 273.29
Chemical Name or Material 2-(1-Naphthylmethyl)-2-imidazoline nitrate
Molecular Formula C14H14N2·HNO3
Melting Point 168° to 170°C
MDL Number MFCD00014316
CAS 5144-52-5
Beilstein 3779329
Synonym Naphazoline nitrate
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99%
Quantity 25g
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
RTECS Number NJ4376000
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.