missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(1-Naphthylmethyl)-2-imidazoline nitrate, 99%

Manufacturer:  Alfa Aesar™ L1261722

 View more versions of this product

Catalog No. AAL1261722



Specifications

Specifications

2-(1-Naphthylmethyl)-2-imidazoline nitrate
100g
C14H15N3O3
3779329
ZAHXYMFVNNUHCP-UHFFFAOYSA-N
2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazole;nitric acid
82332
99%
168°C to 170°C
5144-52-5
MFCD00014316
naphazoline nitrate, privine, naphcon, 2-1-naphthylmethyl-2-imidazoline nitrate, unii-sc99gr1t5s, naphthisen, naphthizen, vistalbalon, imidazyl, naftizin
C1CN=C(N1)CC2=CC=CC3=CC=CC=C32.[N+](=O)(O)[O-]
273.292
273.29

Chemical Identifiers

5144-52-5
273.292
ZAHXYMFVNNUHCP-UHFFFAOYSA-N
82332
C1CN=C(N1)CC2=CC=CC3=CC=CC=C32.[N+](=O)(O)[O-]
C14H15N3O3
MFCD00014316
naphazoline nitrate, privine, naphcon, 2-1-naphthylmethyl-2-imidazoline nitrate, unii-sc99gr1t5s, naphthisen, naphthizen, vistalbalon, imidazyl, naftizin
2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazole;nitric acid
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NJ4376000

Documents