missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(1-Naphthyl)pyrrolidine, 95%

$181.31 - $540.53

Chemical Identifiers

CAS 121193-91-7
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.281
MDL Number MFCD02663484
InChI Key FEJHFYHZVJWOOE-UHFFFAOYSA-N
Synonym 2-1-naphthyl pyrrolidine, 2-naphthalen-1-yl pyrrolidine, 2-naphthalen-1-yl-pyrrolidine, acmc-20ct66, pyrrolidine,2-1-naphthalenyl
PubChem CID 249758
IUPAC Name 2-naphthalen-1-ylpyrrolidine
SMILES C1CC(NC1)C2=CC=CC3=CC=CC=C32
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61800MD View Documents Alfa Aesar™
H61800MD
250mg Each for $181.31
 
AAH6180003 View Documents Alfa Aesar™
H6180003
1g Each for $540.53
 
Description

Description

Applications
2-(1-Naphthyl)pyrrolidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

121193-91-7
197.281
FEJHFYHZVJWOOE-UHFFFAOYSA-N
249758
C1CC(NC1)C2=CC=CC3=CC=CC=C32
C14H15N
MFCD02663484
2-1-naphthyl pyrrolidine, 2-naphthalen-1-yl pyrrolidine, 2-naphthalen-1-yl-pyrrolidine, acmc-20ct66, pyrrolidine,2-1-naphthalenyl
2-naphthalen-1-ylpyrrolidine

Specifications

Yellow
121193-91-7
MFCD02663484
Slightly soluble in water.
C1CC(NC1)C2=CC=CC3=CC=CC=C32
197.281
197.28
95%
250mg
C14H15N
2-1-naphthyl pyrrolidine, 2-naphthalen-1-yl pyrrolidine, 2-naphthalen-1-yl-pyrrolidine, acmc-20ct66, pyrrolidine,2-1-naphthalenyl
FEJHFYHZVJWOOE-UHFFFAOYSA-N
2-naphthalen-1-ylpyrrolidine
249758
Liquid
2-(1-Naphthyl)pyrrolidine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures