Please login to your online account to display your discounted pricing

2-(1-Naphthyl)ethanoyl chloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(1-Naphthyl)ethanoyl chloride
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H9ClO
Formula Weight 204.66
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S07397DA

 
fisher scientific company
S07397DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 204.66
Quantity 1g
Assay 95%
Chemical Name or Material 2-(1-Naphthyl)ethanoyl chloride
Assay Percent Range 95%
CAS 5121-00-6
Molecular Formula C12H9ClO