Please login to your online account to display your discounted pricing

2-(1-Naphthyl)ethanoyl chloride, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  S07397DA

 View more versions of this product

Catalog No. S07397DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 204.66
Quantity 1g
Assay 95%
Assay Percent Range 95%
Chemical Name or Material 2-(1-Naphthyl)ethanoyl chloride
CAS 5121-00-6
Molecular Formula C12H9ClO

Specifications

Chemical Name or Material 2-(1-Naphthyl)ethanoyl chloride
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H9ClO
Formula Weight 204.66