missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(1-Naphthyl)ethanol 95.0+%, TCI America™
$160.92 - $419.73
Chemical Identifiers
CAS | 773-99-9 |
---|---|
Molecular Formula | C12H12O |
Molecular Weight (g/mol) | 172.227 |
MDL Number | MFCD00004050 |
InChI Key | RXWNCMHRJCOWDK-UHFFFAOYSA-N |
Synonym | 1-naphthaleneethanol, 2-1-naphthyl ethanol, 2-naphthalen-1-yl ethanol, 1-2-hydroxyethyl naphthalene, naphthalen-1-ethanol, 2-naphthylethanol, 1-naphthyl ethanol, 1-naphthalene ethanol, 2-naphthalen-1-yl ethan-1-ol, 1-napthaleneethanol |
PubChem CID | 13047 |
IUPAC Name | 2-naphthalen-1-ylethanol |
SMILES | C1=CC=C2C(=C1)C=CC=C2CCO |
Chemical Identifiers
773-99-9 | |
172.227 | |
RXWNCMHRJCOWDK-UHFFFAOYSA-N | |
13047 | |
C1=CC=C2C(=C1)C=CC=C2CCO |
C12H12O | |
MFCD00004050 | |
1-naphthaleneethanol, 2-1-naphthyl ethanol, 2-naphthalen-1-yl ethanol, 1-2-hydroxyethyl naphthalene, naphthalen-1-ethanol, 2-naphthylethanol, 1-naphthyl ethanol, 1-naphthalene ethanol, 2-naphthalen-1-yl ethan-1-ol, 1-napthaleneethanol | |
2-naphthalen-1-ylethanol |
Specifications
773-99-9 | |
White-Yellow | |
C12H12O | |
5 g | |
RXWNCMHRJCOWDK-UHFFFAOYSA-N | |
2-naphthalen-1-ylethanol | |
13047 | |
≥95.0% (GC) | |
2-(1-Naphthyl)ethanol |
60°C | |
179°C | |
MFCD00004050 | |
1-naphthaleneethanol, 2-1-naphthyl ethanol, 2-naphthalen-1-yl ethanol, 1-2-hydroxyethyl naphthalene, naphthalen-1-ethanol, 2-naphthylethanol, 1-naphthyl ethanol, 1-naphthalene ethanol, 2-naphthalen-1-yl ethan-1-ol, 1-napthaleneethanol | |
C1=CC=C2C(=C1)C=CC=C2CCO | |
172.227 | |
172.23 | |
Crystalline Powder |
Safety and Handling
TSCA : No