missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(1-Naphthyl)ethanol, 95%, ACROS Organics™

Chemical Identifiers

CAS 773-99-9
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C12H12O
Linear Formula C10H7CH2CH2OH
Formula Weight 172.23
Synonym 1-(2-Hydroxyethyl)naphthalene
Chemical Name or Material 2-(1-Naphthyl)ethanol
Assay ≥94.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC179600100 SDS Thermo Fisher Scientific
AC179600100
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC179600500 SDS Thermo Fisher Scientific
AC179600500
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

773-99-9
100.0
C12H12O
172.23
2-(1-Naphthyl)ethanol
94.0
95%
C10H7CH2CH2OH
1-(2-Hydroxyethyl)naphthalene
≥94.0%

Specifications

773-99-9
100.0
Solid or Crystalline Powder
C12H12O
172.23
10g
2-(1-Naphthyl)ethanol
94.0
Authentic
95%
C10H7CH2CH2OH
White to off-white
1-(2-Hydroxyethyl)naphthalene
≥94.0%