missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1-Naphthyl)ethanesulfonyl Chloride 97.0+%, TCI America™
$113.71 - $342.98
Chemical Identifiers
| CAS | 104296-63-1 |
|---|---|
| Molecular Formula | C12H11ClO2S |
| Molecular Weight (g/mol) | 254.728 |
| MDL Number | MFCD00191560 |
| InChI Key | IAZQBUVCHCLINF-UHFFFAOYSA-N |
| Synonym | 2-(1-Naphthyl)ethylsulfonyl Chloride |
| PubChem CID | 21901305 |
| IUPAC Name | 2-naphthalen-1-ylethanesulfonyl chloride |
| SMILES | C1=CC=C2C(=C1)C=CC=C2CCS(=O)(=O)Cl |
Chemical Identifiers
| 104296-63-1 | |
| 254.728 | |
| IAZQBUVCHCLINF-UHFFFAOYSA-N | |
| 21901305 | |
| C1=CC=C2C(=C1)C=CC=C2CCS(=O)(=O)Cl |
| C12H11ClO2S | |
| MFCD00191560 | |
| 2-(1-Naphthyl)ethylsulfonyl Chloride | |
| 2-naphthalen-1-ylethanesulfonyl chloride |
Specifications
| 104296-63-1 | |
| C12H11ClO2S | |
| 1 g | |
| 2-(1-Naphthyl)ethylsulfonyl Chloride | |
| C1=CC=C2C(=C1)C=CC=C2CCS(=O)(=O)Cl | |
| 254.728 | |
| 254.73 | |
| Crystalline Powder |
| 50°C | |
| MFCD00191560 | |
| 3261 | |
| IAZQBUVCHCLINF-UHFFFAOYSA-N | |
| 2-naphthalen-1-ylethanesulfonyl chloride | |
| 21901305 | |
| ≥97.0% (HPLC,T) | |
| 2-(1-Naphthyl)ethanesulfonyl Chloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator