Learn More
2-(1-Methoxy)propyl acetate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 413900050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-(1-Methoxy)propyl acetate | |
98.5 | |
0.9600g/mL | |
42°C | |
98.5% min. (GC) | |
C6H12O3 | |
CH3OCH2CH(CH3)OC(O)CH3 | |
5kg | |
0.96 | |
Solubility in water: 19.8g/L (25°C) | |
CC(COC)OC(=O)C | |
132.16 | |
132.16 |
108-65-6 | |
100.0 | |
146.0°C | |
Authentic | |
Plastic drum | |
1.4010 to 1.4030 | |
MFCD00038500 | |
1993 | |
1-methoxy-2-propyl acetate, pgmea, 2-acetoxy-1-methoxypropane, propylene glycol monomethyl ether acetate, propylene glycol methyl ether acetate, 1-methoxy-2-acetoxypropane, 2-propanol, 1-methoxy-, acetate, 2-methoxy-1-methylethyl acetate, propyleneglycol monomethyl ether acetate, acetic acid, 2-methoxy-1-methylethyl ester | |
LLHKCFNBLRBOGN-UHFFFAOYSA-N | |
1-methoxypropan-2-yl acetate | |
7946 | |
99% |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge
GHS Signal Word: Warning
EINECSNumber : 203-603-9
RUO â Research Use Only