missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione, TCI America™
$137.50 - $412.52
Chemical Identifiers
CAS | 172611-72-2 |
---|---|
Molecular Formula | C13H20O3 |
Molecular Weight (g/mol) | 224.3 |
MDL Number | MFCD01862890 |
InChI Key | HNPWTDUZIXAJSA-UHFFFAOYSA-N |
Synonym | 2-(1-Hydroxy-3-methylbutylidene)dimedone |
PubChem CID | 22728397 |
IUPAC Name | 5,5-dimethyl-2-(3-methylbutanoyl)cyclohexane-1,3-dione |
SMILES | CC(C)CC(=O)C1C(=O)CC(CC1=O)(C)C |
Chemical Identifiers
172611-72-2 | |
224.3 | |
HNPWTDUZIXAJSA-UHFFFAOYSA-N | |
22728397 | |
CC(C)CC(=O)C1C(=O)CC(CC1=O)(C)C |
C13H20O3 | |
MFCD01862890 | |
2-(1-Hydroxy-3-methylbutylidene)dimedone | |
5,5-dimethyl-2-(3-methylbutanoyl)cyclohexane-1,3-dione |
Specifications
Yellow | |
172611-72-2 | |
MFCD01862890 | |
HNPWTDUZIXAJSA-UHFFFAOYSA-N | |
5,5-dimethyl-2-(3-methylbutanoyl)cyclohexane-1,3-dione | |
22728397 | |
Clear Liquid at 20°C | |
2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione |
1g | |
C13H20O3 | |
2-(1-Hydroxy-3-methylbutylidene)dimedone | |
CC(C)CC(=O)C1C(=O)CC(CC1=O)(C)C | |
224.3 | |
224.30 | |
≥98.0% (GC) |
Safety and Handling
TSCA : No