Please login to your online account to display your discounted pricing

2-(1-Benzothiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 95%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC24839DA

 View more versions of this product

Catalog No. CC24839DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 260.16
Quantity 1g
Assay 95%
CAS Max % 100.0
Synonym 2-(1-Benzothiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane (IUPAC)
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD05664405
Molecular Formula C14H17BO2S
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
CAS Min % 94.5
Assay Percent Range 95%
Chemical Name or Material 2-(1-Benzothiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CAS 501945-71-7

Specifications

Chemical Name or Material 2-(1-Benzothiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CAS 501945-71-7
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C14H17BO2S
Formula Weight 260.16
MDL Number MFCD05664405
Synonym 2-(1-Benzothiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane (IUPAC)
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents